Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is Hela cell lysate.
PA5-34348 can be used with blocking peptide PEP-1391.
YPEL2 (yippee-like 2) belongs to a family of five yippee-like proteins, all of which localize to the centrosome or mitotic spindle and are widely expressed in both adult and fetal tissue. This localization plus the fact that the family of human YPEL proteins share a high degree of sequence homology across species suggests that these proteins may have a conserved function involved in cell division. YPEL2 might be an important factor during the development and malignant transformation of tissues, most notably pancreatic and breast tumors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DiGeorge syndrome-related protein; ortholog of human yippee-like 2 YPEL2; Protein yippee-like 2; yippee-like 2
Gene Aliases: 6430570G24; E130113K08Rik; FKSG4; YPEL2
UniProt ID: (Human) Q96QA6, (Mouse) Q65Z95
Entrez Gene ID: (Human) 388403, (Mouse) 77864
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support