Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKKPSAKQQ KEVEKVKPQC KEVHQTLILD PAQRKRLQQQ MQQHVQLLTQ IHLLATCNPN LNPEASSTRI CLKELGTFAQ SSIALHHQYN PKFQTLFQPC NLMGAMQLIE DFSTHVSIDC SPHKTVKKTA NEFPCLPKQV AWILATSKVF MYPELLPVCS LKAKNPQDKI LFTKAEDNLL ALGLKHFEGT EFLNPLISKY LLTCKTARQL TVRIKNLNMN RAPDNIIKFY KKTKQLPVLG KCCEEIQPHQ WKPPIEREEH RLPFWLKASL PSIQEELRHM ADGAREVGNM TGTTEINSDQ GLEKDNSELG SETRYPLLLP KGVVLKLKPV ADRFPKKAWR QKRSSVLKPL LIQPSPSLQP SFNPGKTPAQ STHSEAPPSK MVLRIPHPIQ PATVLQTVPG V (1-400 aa encoded by BC003500)
The encoded gene product presumably interacts with YY1 protein; however, its exact function is not known. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hepatocellular carcinoma susceptibility protein; Hepatocellular carcinoma-associated protein 2; YY1-associated protein 1
Gene Aliases: HCCA1; HCCA2; YAP; YY1AP; YY1AP1
UniProt ID: (Human) Q9H869
Entrez Gene ID: (Human) 55249
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support