Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: MGFSNMEDDG PEEEERVAEP QANFNTPQAL RFEELLANLL NEQHQIAKEL FEQLKMKKPS AKQQKEVEKV KPQCKEVHQT LILDPAQRKR LQQQMQQHVQ LLTQIHLLAT CNPNLNPEAS STRICLKELG TFAQSSIALH HQYNPKFQTL FQPCNLMGAM QLIEDFSTHV SIDCSPHKTV KKTANEFPCL PKQVAWILAT SKVFMYPELL PVCSLKAKNP QDKILFTKAE DNLLALGLKH FEGTEFLNPL ISKYLLTCKT ARQLTVRIKN LNMNRAPDNI IKFYKKTKQL PVLGKCCEEI QPHQWKPPIE REEHRLPFWL KASLPSIQEE LRHMADGARE VGNMTGTTEI NSDQGLEKDN SELGSETRYP LLLPKGVVLK LKPVADRFPK KAWRQKRSSV LKPLLIQPSP SLQPSFNPGK TPAQSTHSEA PPSKMVLRIP HPIQPATVLQ TVPGVPPLGV SGGESFESPA ALPAMPPEAR TSFPLSESQT LLSSALVPKV MMPSPASSMF RKPYVRRRPS KRRGARAFRC IKPAPVIHPA SVIFTVPATT VKIVSLGGGC NMIQPVNAAV AQSPQTIPIA TLLVNPTSFP CPLNQPLVAS SVSPLIVSGN SVNLPIPSTP EDKAHMNVDI ACAVADGENA FQGLEPKLEP QELSPLSATV FPKVEHSPGP PPVDKQCQEG LSENSAYRWT VVKTEEGRQA LEPLPQGIQE SLNNSSPGDL EEVVKMEPED ATEEISGFL
The encoded gene product presumably interacts with YY1 protein; however, its exact function is not known. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hepatocellular carcinoma susceptibility protein; Hepatocellular carcinoma-associated protein 2; YY1-associated protein 1
Gene Aliases: HCCA1; HCCA2; YAP; YY1AP; YY1AP1
UniProt ID: (Human) Q8IXA6
Entrez Gene ID: (Human) 55249
Molecular Function: transcription cofactor gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support