Search Thermo Fisher Scientific
Progen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
2D8 specifically reacts with Yersinia enterocolitica serogroup O:3 strains in both agglutination and immunofluorescence assays. The antibody does not react with Y. enterocolitica O:9 and O:8 serogroups.
Yersinia enterocolitica serotype O:3 is a Gram-negative, bacillus-shaped bacterium, belonging to the family Yersiniaceae. It is motile at temperatures of 22-29 degrees celsius, but becomes nonmotile at normal human body temperature. Yersinia enterocolitica is thought to be a significant food-borne pathogen, although pathogenic isolates have seldom been isolated from foods, except from edible pig offal. In case control studies, a correlation has been demonstrated between the consumption of raw or undercooked pork and yersiniosis. Genotypes of Y. enterocolitica strains found in humans, further supporting the association between yersiniosis and consumption of pork.