Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: KEKPPDQKLV VRSSRDNYVL TQCDFEDDAK PLCDWSQVSA DDEDWVRASG PSPTGSTGAP GGYPNGEGSY LHMESNSFHR GGVARLLSPD LWEQG
This gene encodes a sperm membrane protein that binds the zona pellucida of the egg in a species-specific manner. The encoded protein may be involved in signaling or gamete recognition. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zonadhesin
Gene Aliases: ZAN
UniProt ID: (Human) Q9Y493
Entrez Gene ID: (Human) 7455
Molecular Function: extracellular matrix protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support