Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BRC1744; Zinc finger protein 13; Zinc finger protein 45; zinc finger protein 45 (a Kruppel-associated box (KRAB) domain polypeptide); Zinc finger protein KOX5
Gene Aliases: KOX5; ZNF13; ZNF45
UniProt ID: (Human) P17016
Entrez Gene ID: (Human) 7596
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support