Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 6
Recommended positive controls: 293T, NIH-3T3, JC, BCL-1.
Predicted reactivity: Mouse (100%), Rat (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene, which was initially designated as dendritic cell-derived BTB/POZ zinc finger (DPZF), belongs to a family of transcription factors with an N-terminal BTB/POZ domain and a C-terminal DNA-bindng zinc finger domain. The BTB/POZ domain is a hydrophobic region of approximately 120 aa which mediates association with other BTB/POZ domain-containing proteins. This gene acts as a transcriptional repressor and plays a role in many processes including neurogenesis, glucose homeostasis, and postnatal growth. Mutations in this gene have been associated with Primrose syndrome as well as the 3q13.31 microdeletion syndrome. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq, Feb 2017]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BTB/POZ domain zinc finger factor HOF; BTB/POZ zinc finger protein DPZF; dendritic cell-derived BTB/POZ zinc finger; Dendritic-derived BTB/POZ zinc finger protein; POZ/zinc finger transcription factor ODA-8; Zinc finger and BTB domain-containing protein 20; Zinc finger protein 288
Gene Aliases: 1300017A20Rik; 7330412A13Rik; A930017C21Rik; D16Wsu73e; DPZF; HOF; ODA-8S; Oda8; PRIMS; RGD1560387; ZBTB20; Zfp288; ZNF288
UniProt ID: (Human) Q9HC78, (Mouse) Q8K0L9
Entrez Gene ID: (Human) 26137, (Rat) 288105, (Mouse) 56490
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support