Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEPSPLSPSG AALPLPLSLA PPPLPLPAAA VVHVSFPEVT SALLESLNQQ RLQGQLCDVS IRVQGREFRA HRAVLAASSP YFHDQVLLKG MTSISLPSVM DPGAFETVLA SAYTGRLSMA AADIVNFLTV GSVLQMWHIV DKCTELLREG RASATTTITT AAATSVTVPG AGVPSGSGGT VAPATMGSAR SHASSRASEN QSPSSSNYFS PRESTDFSSS SQEAFAASAV GSGERRGGGP VFPAPVVGSG GATSGKLLLE ADELCDDGGD GRGAVVPGAG LRRPTYTPPS IMPQKHWVYV KRGGNCPAPA PLVPQDPDLE EEEEEEDLVL TCEDDEDEEL GGSSRVPVGG GPEATLSISD VRTLSEPPDK GEEQVNFCES SNDFGPYEGG GPVAGLDDSG GPTPSSYAPS HPPRPLLPLD MQGNQILVFP SSSSSSSSQA PGQPPGNQAE HGAVTVGGTS VGSLGVPGSV GGVPGGTGSG DGNKIFLCHC GKAFSHKSMR DRHVNMHLNL RPFDCPVCNK KFKMKHHLTE HMKTHTGLKP YECGVCAKKF MWRDSFMRHR GHCERRHRLG GVGAVPGPGT PTGPSLPSKR ESPGVGGGSG DEASAATPPS SRRVWSPPRV HKVEMGFGGG GGAN
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein BING1; Zinc finger and BTB domain-containing protein 22; Zinc finger and BTB domain-containing protein 22A; Zinc finger protein 297
Gene Aliases: BING1; fru; fruitless; ZBTB22; ZBTB22A; ZNF297; ZNF297A
UniProt ID: (Human) O15209
Entrez Gene ID: (Human) 9278
Molecular Function: non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support