Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Additional Information: This antibody is expected to recognize both reported isoforms (NP_055612.2; NP_001157785.1).
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
May be involved in BMP2-induced transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: POZ (BTB) and AT hook containing zinc finger 2; Zinc finger and BTB domain-containing protein 24; Zinc finger protein 450
Gene Aliases: BIF1; ICF2; KIAA0441; PATZ2; ZBTB24; ZNF450
UniProt ID: (Human) O43167
Entrez Gene ID: (Human) 9841
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support