Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: DTEPVELNCN FSFSRKRKMS CTICGHKFPR KSQLLEHMYT HKGKSYRYNR CQRFGNALAQ RFQPYCDSWS DVSLKSSRLS QEHLDLPCAL ESELTQENVD TILVE
May be involved in transcriptional regulation. Classified as a putative transcription factor; human homolog is a DNA binding protein with two distantly spaced zinc fingers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger and BTB domain-containing protein 25; Zinc finger protein 46; Zinc finger protein KUP
Gene Aliases: C14orf51; KUP; ZBTB25; ZNF46
UniProt ID: (Human) P24278
Entrez Gene ID: (Human) 7597
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support