Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: DTEPVELNCN FSFSRKRKMS CTICGHKFPR KSQLLEHMYT HKGKSYRYNR CQRFGNALAQ RFQPYCDSWS DVSLKSSRLS QEHLDLPCAL ESELTQENVD TILVE
May be involved in transcriptional regulation. Classified as a putative transcription factor; human homolog is a DNA binding protein with two distantly spaced zinc fingers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger and BTB domain-containing protein 25; Zinc finger protein 46; Zinc finger protein KUP
Gene Aliases: C14orf51; KUP; ZBTB25; ZNF46
UniProt ID: (Human) P24278
Entrez Gene ID: (Human) 7597
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support