Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger and BTB domain-containing protein 26; Zinc finger protein 481; zinc finger protein 483; Zinc finger protein Bioref
Gene Aliases: A630026F21Rik; bioref; KIAA1572; ZBTB26; ZNF481
UniProt ID: (Human) Q9HCK0
Entrez Gene ID: (Human) 57684, (Rat) 311910, (Mouse) 320633
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support