Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLREFSKWGV EASPGKAWER KRSLLRGAVG RYRGATGGDL FWAPFPSWGT MEFPEHSQQL LQSLREQRSQ GFLCDCTVMV GSTQFLAHRA VLASCSPFFQ LFYKERELDK RDLVCIHNEI VTAPAFGLLL DFMYAGQLTL RGDTPVEDVL AAASYLHMND IVKVCKRRLQ ARALAEADST KKEEETNSQL PSLEFLSSTS RGTQPSLASA ETSGHWGKGE WKGSAAPSPT VRPPDEPPMS SGADTTQPGM EVDAPHLRAP HPPVADVSLA SPSSSTETIP TNYFSSGISA VSLEPLPSLD VGPESLRVVE PKDPGGPLQG FYPPASAPTS APAPVSAPVP SQAPAPAEAE LVQVKVEAIV ISDEETDVSD EQPQGPERAF PSGGAVYGAQ PSQPEAFEDP GAAGLEEVGP SDHFLPTDPH LPYHLLPGAG QYHRGLVTSP LPAPASLHEP LYLSSEYEAA PGSFGVFTED VPTCKTCGKT FSCSYTLRRH ATVHTRERPY ECRYCLRSYT QSGDLYRHIR KAHNEDLAKR SKPDPEVGPL LGVQPLPGSP TADRQSSSGG GPPKDFVLAP KTNI
The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. While little is known about ZBTB3, the related protein ZBTB2 is thought to be phosphorylated in response to the DNA damage, probably by either ATM or ATR. Other ZBTB proteins, such as ZBTB4 and ZBTB38 bind methylated DNA and repress transcription, suggesting that ZBTB3 may also act as a transcription repressor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Entrez Gene ID: (Human) 79842
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support