Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
FIGURE: 1 / 1
DNA-binding protein that binds to the to a 5'-TGTACAGTGT-3' core sequence. May function as a transcriptional transactivator and transcriptional repressor. Probably exerts its repressor effect by preventing GATA3 from binding to DNA. May play a role in regulating the differentiation and activation of helper T-cells. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FANCC-interacting protein; Fanconi anemia zinc finger protein; OTTHUMP00000045790; repressor of GATA; Testis zinc finger protein; Zinc finger and BTB domain-containing protein 32; Zinc finger protein 538
Gene Aliases: FAXF; FAZF; Rog; TZFP; ZBTB32; ZNF538
UniProt ID: (Human) Q9Y2Y4
Entrez Gene ID: (Human) 27033
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support