Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ELAEFEIGA SSSSSTEALI SAVMAGRSIS PWLARRTSPA NSSGDSAIAS CHDGGSSYGK EDQEPKADGP DDVSSQPLWP GDVGYGPLRI KEEQVSPSQY GGSELPSAKD GAVQNSFSEQ SAGDAWQPTG RRKNRKNKET VRHITQQVED DSRASSPVPS FLPTSGWPFS SRDSNADLSV TEASSSDSRG ERAELYAQVE EGLLGGEASY LGPPLTPEKD DALHQATAVA NLRAALMSKN SLLSLKADVL GDDGSLLFEY LPRGAHSLSL NEFTVIRK (140-416 aa encoded by BC073800)
ZBTB46 (Zinc Finger And BTB Domain Containing 46) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include nucleic acid binding. ZBTB46 also functions as a transcriptional repressor for PRDM1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BTB (POZ) domain containing 4; BTB-ZF protein expressed in effector lymphocytes; BTB/POZ domain-containing protein 4; BZEL; Zinc finger and BTB domain-containing protein 46; Zinc finger protein 340
Gene Aliases: 2610019F01Rik; 4933406L05Rik; BTBD4; BZEL; dJ583P15.7; dJ583P15.8; RINZF; ZBTB46; ZNF340
UniProt ID: (Human) Q86UZ6, (Mouse) Q8BID6
Entrez Gene ID: (Human) 140685, (Rat) 311718, (Mouse) 72147
Molecular Function: non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support