Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MNNRKEDMEI ASHYRHLLRE LNEQRQHGVL CDVCVVVEGK VFKAHKNVLL GSSRYFKTLY CQVQKTSEQA TVTHLDIVTA QGFKAIIDFM YSAHLALTSR NVIEVMSAAS FLQMTDIVQA CHDFIKAALD ISIKSDASDE LAEFEIGASS SSSTEALISA VMAGRSISPW LARRTSPANS SGDSAIASCH DGGSSYGKED QEPKADGPDD VSSQPLWPGD VGYGPLRIKE EQVSPSQYGG SELPSAKDGA VQNSFSEQSA GDAWQPTGRR KNRKNKETVR HITQQVEDDS RASSPVPSFL PTSGWPFSSR DSNADLSVTE ASSSDSRGER AELYAQVEEG LLGGEASYLG PPLTPEKDDA LHQATAVANL RAALMSKNSL LSLKADVLGD DGSLLFEYLP RGAHSLSLNE FTVIRKKFKC PYCSFSAMHQ CILKRHMRSH TGERPYPCEI CGKKFTRREH MKRHTLVHSK DKKYVCKVCS RVFMSAASVG IRHGSRRHGV CTDCAGRGMA GPLDHGGGGG EGSPEALFPG DGPYLEDPED PRGEAEELGE DDEGLAPEDA LLADDKDEED SPRPRSPPGG PDKDFAWLS
ZBTB46 (Zinc Finger And BTB Domain Containing 46) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include nucleic acid binding. ZBTB46 also functions as a transcriptional repressor for PRDM1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BTB (POZ) domain containing 4; BTB-ZF protein expressed in effector lymphocytes; BTB/POZ domain-containing protein 4; BZEL; Zinc finger and BTB domain-containing protein 46; Zinc finger protein 340
Gene Aliases: BTBD4; BZEL; dJ583P15.7; dJ583P15.8; RINZF; ZBTB46; ZNF340
UniProt ID: (Human) Q86UZ6
Entrez Gene ID: (Human) 140685
Molecular Function: non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support