Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDGSFVQHSV RVLQELNKQR EKGQYCDATL DVGGLVFKAH WSVLACCSHF FQSLYGDGSG GSVVLPAGFA EIFGLLLDFF YTGHLALTSG NRDQVLLAAR ELRVPEAVEL CQSFKPKTSV GQAAGGQSGL GPPASQNVNS HVKEPAGLEE EEVSRTLGLV PRDQEPRGSH SPQRPQLHSP AQSEGPSSLC GKLKQALKPC PLEDKKPEDC KVPPRPLEAE GAQLQGGSNE WEVVVQVEDD GDGDYMSEPE AVLTRRKSNV IRKPCAAEPA LSAGSLAAEP AENRKGTAVP VECPTCHKKF LSKYYLKVHN RKHTGEKPFE CPKCGKCYFR KENLLEHEAR NCMNRSEQVF TCSVCQETFR RRMELRVHMV SHTGEMPYKC SSCSQQFMQK KDLQSHMIKL HGAPKPHACP TCAKCFLSRT ELQLHEAFKH RGEKLFVCEE CGHRASSRNG LQMHIKAKHR NERPHVCEFC SHAFTQKANL NMHLRTHTGE KPFQCHLCGK TFRTQASLDK HNRTHTGERP FSCEFCEQRF TEKGPLLRHV ASRHQEGRPH FCQICGKTFK AVEQLRVHVR RHKGVRKFEC TECGYKFTRQ AHLRRHMEIH DRVENYNPRQ RKLRNLIIED EKMVVVALQP PAELEVGSAE VIVESLAQGG LASQLPGQRL CAEESFTGPG VLEPSLIITA AVPEDCDT
ZBTB48 contains 1 BTB (POZ) domain and 11 C2H2-type zinc fingers. It belongs to the krueppel C2H2-type zinc-finger protein family and binds to and regulates the J and/or S elements in MHC II promoter.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GLI-Kruppel family member HKR3; hKR3; Krueppel-related zinc finger protein 3; Telomere zinc finger-associated protein; TZAP; Zinc finger and BTB domain-containing protein 48; Zinc finger protein 855
Gene Aliases: HKR3; pp9964; TZAP; ZBTB48; ZNF855
UniProt ID: (Human) P10074
Entrez Gene ID: (Human) 3104
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support