Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SQPSEVTHYK SSKREVRTSD SSSHVSQSEE QAQIDAEMDS TPVGYQYGQG SDVTSKSFPD DLPRMRFKCP YC
Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 67%.
The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. ZBTB8, also known as BOZ-F1, is present in multiple tissues, and is highly expressed in the cytoplasm of the placenta. While little is known about ZBTB8, other ZBTB proteins, such as ZBTB4 bind methylated DNA and repress transcription. Another ZBTB proteins, ZBTB7A, has been implicated as a proto-oncogene whose overexpression contributes to malignancy in breast cancer, suggesting that ZBTB8 may act as a transcriptional repressor or be involved in tumorigenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BOZ-F1; BTB/POZ and zinc-finger domain-containing factor; BTB/POZ and zinc-finger domains factor on chromosome 1; zinc finger and BTB domain containing 8; Zinc finger and BTB domain-containing protein 8A
Gene Aliases: BOZF1; ZBTB8; ZBTB8A; ZNF916A
UniProt ID: (Human) Q96BR9
Entrez Gene ID: (Human) 653121
Molecular Function: non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support