Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Dog: 93%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Mouse: 79%; Rabbit: 86%; Rat: 86%
ZBTB8B may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: putative zinc finger and BTB domain-containing protein 8B; Zinc finger and BTB domain-containing protein 8B
Gene Aliases: ZBTB8B; ZNF916B
UniProt ID: (Human) Q8NAP8
Entrez Gene ID: (Human) 728116
Molecular Function: non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support