Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MEHPSKMEFF QKLGYDREDV LRVLGKLGEG ALVNDVLQEL IRTGSRPGAL EHPAAPRLVP RGSCGVPDSA QRGPGTALEE DFRTLASSLR
ZC3H12D, also known as MCPIP4, is a member of a family of novel CCCH-zinc finger proteins that includes ZC3H12A, a protein that is thought to be involved in macrophage activation, host immunity and inflammatory diseases. ZC3H12D is highly expressed in spleen, intestine, and colon. Similar to ZC3H12A and ZC3H12C, ZC3H12D expression in murine macrophages is also increased after treatment with lipopolysaccharide (LPS), although not to the same level as ZC3H12A. This pattern of expression was not duplicated in human-derived macrophages, suggesting it also may play a different role in host immunity and inflammatory response.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MCP-induced protein 4; p34; Probable ribonuclease ZC3H12D; RP1-281H8.5; Transformed follicular lymphoma; tumor suppressor TFL; Zinc finger CCCH domain-containing protein 12D
Gene Aliases: C6orf95; dJ281H8.1; MCPIP4; p34; TFL; ZC3H12D
UniProt ID: (Human) A2A288
Entrez Gene ID: (Human) 340152
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support