Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSSKDFFACG HSGHWARGCP RGGAGGRRGG GHGRGSQCGS TTLSYTCYCC GESGRNAKNC VLLGNICYNC GRSGHIAKDC KDPKRERRQH CYTCGRLGHL ARDCDRQKEQ KCYSCGKLGH IQKDCAQVKC YRCGEIGHVA INCSKARPGQ LLPLRQIPTS SQGMSQ
This gene appears to represent an intronless retrocopy of a related multi-exon gene located on chromosome 3. However, the CDS of this intronless gene remains relatively intact, it is conserved in other mammalian species, it is known to be transcribed, and it is therefore thought to encode a functional protein. The encoded protein contains six CCHC-type zinc fingers, and is thus thought to function as a transcription factor. [provided by RefSeq, May 2010].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger CCHC domain-containing protein 13; zinc finger, CCHC domain containing 13
Gene Aliases: CNBP2; ZCCHC13; ZNF9L
UniProt ID: (Human) Q8WW36
Entrez Gene ID: (Human) 389874
Molecular Function: nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support