Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Positive Samples: HeLa, MCF7, SKOV3, A-549, Mouse brain, Mouse lung, Mouse testis, Rat brain, Rat lung
Immunogen sequence: VFGDEKKYWL LPIFSSLGDG CSFPTRLVGM DPEQASVTNQ NEYARSSGSN QPFPIKPLSE SKNRLLDSES QWLENGAEEG IVKSGTNNHV TVAIEN
Catalyzes palmitoylation of Cys residues on target proteins (PubMed:27153536, PubMed:29326245). Catalyzes palmitoylation of Cys residues in the cytoplasmic C-terminus of EGFR, and modulates the duration of EGFR signaling by modulating palmitoylation-dependent EGFR internalization and degradation (PubMed:27153536). Has a preference for acyl-CoA with C16 fatty acid chains (PubMed:29326245). Can also utilize acyl-CoA with C14 and C18 fatty acid chains (PubMed:29326245). [UniProt]
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acyltransferase ZDHHC20; DHHC domain-containing cysteine-rich protein 20; DHHC-20; DHHC-containing protein 20; DHHC20; membrane-associated DHHC24 zinc finger protein; Palmitoyltransferase ZDHHC20; probable palmitoyltransferase ZDHHC20; Zinc finger DHHC domain-containing protein 20; zinc finger, DHHC domain containing 20; zinc finger, DHHC-type containing 20
Gene Aliases: 4933421L13Rik; 5033406L14Rik; AI448102; B230110O18Rik; DHHC-20; HK11; RGD1305755; ZDHHC20
UniProt ID: (Human) Q5W0Z9, (Mouse) Q5Y5T1
Entrez Gene ID: (Human) 253832, (Rat) 305923, (Mouse) 75965
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support