Search Thermo Fisher Scientific
Immunogen sequence: EDVNLMMSSG QILGIRFLDK LCSQKRDVSS PDENNGQSKS KYCLKINRPI GDAADTQDDD VEEIGDQDDS PSDDTVEGTP PSQEDGKSPT TTLRVQEAIL KELGSEEVRK VNCYGQEVES METPESKDLG SQTPQALTFN DGMSEVKDEQ TPGWTTAASD MKFEYLLYGH HREQIACQAC GKTFSDEGRL RKHEKLHTAD RPFVCEMCTK GFTTQAHLKE HLKIHTGYKP YSCEVCGKSF IRAPDLKKHE RVHSNERPFA CHMCDKAFKH KSHLKDHERR HRGEKPFVCG SCTKAFAKAS DLKRHENNMH SERKQVTPSA IQSETEQLQA AAMAAEAEQQ LETIACS
Transcriptional activator of the dopamine transporter (DAT), binding it's promoter at the consensus sequence 5'-CCTGCACAGTTCACGGA-3'. Binds to 5'-d(GCC)(n)-3' trinucleotide repeats in promoter regions and acts as a repressor of the FMR1 gene. Transcriptional repressor of MYC and thymidine kinase promoters.
仅用于科研。不用于诊断过程。未经明确授权不得转售。
蛋白别名: ZF5; Zfp-161; ZFP161 zinc finger protein; Zinc finger and BTB domain-containing protein 14; Zinc finger protein 161 homolog; Zinc finger protein 478; Zinc finger protein 5 homolog; zinc finger protein homologous to Zfp161 in mouse
基因别名: ZBTB14; ZF5; ZFP-161; ZFP-5; ZFP161; ZNF478
UniProt ID: (Human) O43829
Entrez Gene ID: (Human) 7541