Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: EDVNLMMSSG QILGIRFLDK LCSQKRDVSS PDENNGQSKS KYCLKINRPI GDAADTQDDD VEEIGDQDDS PSDDTVEGTP PSQEDGKSPT TTLRVQEAIL KELGSEEVRK VNCYGQEVES METPESKDLG SQTPQALTFN DGMSEVKDEQ TPGWTTAASD MKFEYLLYGH HREQIACQAC GKTFSDEGRL RKHEKLHTAD RPFVCEMCTK GFTTQAHLKE HLKIHTGYKP YSCEVCGKSF IRAPDLKKHE RVHSNERPFA CHMCDKAFKH KSHLKDHERR HRGEKPFVCG SCTKAFAKAS DLKRHENNMH SERKQVTPSA IQSETEQLQA AAMAAEAEQQ LETIACS
Transcriptional activator of the dopamine transporter (DAT), binding it's promoter at the consensus sequence 5'-CCTGCACAGTTCACGGA-3'. Binds to 5'-d(GCC)(n)-3' trinucleotide repeats in promoter regions and acts as a repressor of the FMR1 gene. Transcriptional repressor of MYC and thymidine kinase promoters.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ZF5; Zfp-161; ZFP161 zinc finger protein; Zinc finger and BTB domain-containing protein 14; Zinc finger protein 161 homolog; Zinc finger protein 478; Zinc finger protein 5 homolog; zinc finger protein homologous to Zfp161 in mouse
Gene Aliases: ZBTB14; ZF5; ZFP-161; ZFP-5; ZFP161; ZNF478
UniProt ID: (Human) O43829
Entrez Gene ID: (Human) 7541
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support