Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRGAASASVH EPTPLPGRGA PRTKPRAGRG PTVGTPATLA LPARGRPRSR NGLASKGQRG AAPTGPGHRA LPSRDTALPQ ERNKKLEAVG TGIEPKAMSQ GLVTFGDVAV DFSQEEWEWL NPIQRNLYRK VMLENYRNLA SLGLCVSKPD VISSLEQGKE PWTVKRKMTR AWCPDLKAVW KIKELPLKKD FCEGKLSQAV ITERLTSYNL EYSLLGEHWD YDALFETQPG LVTIKNLAVD FRQQLHPAQK NFCKNGIWEN NSDLGSAGHC VAKPDLVSLL EQEKEPWMVK RELTGSLFSG ECRRAWCGKS TARESGAFTK KAAFSLIYSR KLYPGSLGLE RKLNSGSSK
ZFP28 may be involved in transcriptional regulation. may have a role in embryonic development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Krueppel-like zinc finger factor X6; Zfp-28; zinc finger factor X6; Zinc finger protein 28 homolog
Gene Aliases: KIAA1431; mkr5; ZFP28
UniProt ID: (Human) Q8NHY6
Entrez Gene ID: (Human) 140612
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support