Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Immunogen sequence: MTTTLVSAT IFDLSEVLCK GNKMLNYSAP SAGGCLLDRK AVGTPAGGGF PRRHSVTLPS SKFHQNQLLS SLKGEPAPAL SSRDSRFRDR SFSEGGERLL PTQKQPGGGQ VNSSRYKTEL CRPFEENGAC KYGDKCQFAH GIHELRSLTR HPKYKTELCR TFHTIGFCPY GPRCHFIHNA EERRALAGAR DLSADRPRLQ HSFSFAGFPS AAATAAATGL LDSPTSITPP PILSADDLLG SPTLPDGTNN PFAFSSQELA SLFAPSMGLP GGGSPTTFLF RPMSESPHMF DSPPSPQDSL SDQEGYLSSS SSSHSGSDSP TLDNSRRLPI FSRLSISDD (1-338 aa encoded by BC018340 )
This gene is a member of the TIS11 family of early response genes. Family members are induced by various agonists such as the phorbol ester TPA and the polypeptide mitogen EGF. The gene is well conserved across species and has a promoter that contains motifs seen in other early-response genes. The encoded protein contains a distinguishing putative zinc finger domain with a repeating cys-his motif. This putative nuclear transcription factor most likely functions in regulating the response to growth factors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Butyrate response factor 1; early response factor Berg36; EGF-inducible protein CMG1; EGF-response factor 1; ERF-1; mRNA decay activator protein ZFP36L1; TPA-induced sequence 11b; ZFP36 ring finger protein-like 1; ZFP36-like 1; zinc finger protein 36, C3H type-like 1; Zinc finger protein 36, C3H1 type-like 1; zinc finger protein, C3H type, 36-like 1
Gene Aliases: AW742437; AW743212; BERG36; BRF1; Cmg1; D530020L18Rik; ERF-1; ERF1; RNF162B; TIS11B; ZFP36L1
UniProt ID: (Human) Q07352, (Rat) P17431, (Mouse) P23950
Entrez Gene ID: (Human) 677, (Rat) 29344, (Mouse) 12192
Molecular Function:
RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support