Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TIS11D, also known as ZFP36LA (zinc finger protein 36, C3H type-like 2), BRF2 (butyrate response factor 2), ERF2 or RNF162C, is a 494 amino acid protein that localizes to the nucleus and contains two CSH1-type zinc fingers. Belonging to the TIS11 family of early response proteins, TIS11D is thought to function as a nuclear transcription factor that binds to 5'UUAUUUAUUU-3' core RNA sequences and may regulate growth factor-induced cellular responses. The gene encoding TIS11D maps to human chromosome 2, which houses over 1, 400 genes and comprises nearly 8% of the human genome. Harlequin icthyosis, a rare and morbid skin deformity, is associated with mutations in the ABCA12 gene, while the lipid metabolic disorder sitosterolemia is associated with defects in the ABCG5 and ABCG8 genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Butyrate response factor 2; EGF-response factor 2; ERF-2; mRNA decay activator protein ZFP36L2; Protein TIS11D; TISD; TPA-induced sequence 11d; ZFP36 ring finger protein-like 2; ZFP36-like 2; zinc finger protein 36, C3H type-like 1; zinc finger protein 36, C3H type-like 2; zinc finger protein 36, C3H type-like 2-like; Zinc finger protein 36, C3H1 type-like 2; zinc finger protein, C3H type, 36-like 2
Gene Aliases: BRF2; ERF-2; ERF2; RNF162C; TIS11D; ZFP36L2
UniProt ID: (Human) P47974, (Mouse) P23949
Entrez Gene ID: (Human) 678, (Mouse) 12193, (Rat) 298765
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support