Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MDSYFKAAVS DLDKLLDDFE QNPDEQDYLQ DVQNAYDSNH CSVSSELASS QRTSLLPKDQ ECVNSCASSE TSYGTNESSL NEKTLKGLTS IQNEKNVTGL
The ZFYVE16 gene encodes endofin, an endosomal protein implicated in regulating membrane trafficking. It is characterized by the presence of a phosphatidylinositol 3-phosphate-binding FYVE domain positioned in the middle of the molecule.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endofin; Endosome-associated FYVE domain protein; endosome-associated FYVE-domain protein; protein phosphatase 1, regulatory subunit 69; Zinc finger FYVE domain-containing protein 16; zinc finger, FYVE domain containing 16
Gene Aliases: KIAA0305; PPP1R69; ZFYVE16
UniProt ID: (Human) Q7Z3T8
Entrez Gene ID: (Human) 9765
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support