Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a zinc finger transcription factor. The encoded protein likely plays a role in transcriptional repression of interleukin 2. Mutations in this gene have been associated with posterior polymorphous corneal dystrophy-3 and late-onset Fuchs endothelial corneal dystrophy. Alternatively spliced transcript variants encoding different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 110 kDa protein; FLJ34993; FYVE finger-containing Rab5 effector protein rabenosyn-5; MGC126210; RAB effector RBSN; Rabenosyn 5; Rabenosyn-5; Zinc finger FYVE domain-containing protein 20; zinc finger, FYVE domain containing 20
Gene Aliases: 5330426D11Rik; Rabenosyn-5; RBSN; ZFYVE20
UniProt ID: (Human) Q9H1K0, (Mouse) Q80Y56
Entrez Gene ID: (Human) 64145, (Mouse) 78287
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support