Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Recommended positive controls: NT2D1, PC-3, U87-MG, SK-N-SH.
Predicted reactivity: Mouse (86%), Rat (83%), Bovine (88%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein with several transmembrane domains, a Rab11-binding domain and a lipid-binding FYVE finger domain. The encoded protein appears to promote neurite formation. A mutation in this gene has been reported to be associated with hereditary spastic paraplegia, however the pathogenicity of the mutation, which may simply represent a polymorphism, is unclear.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protrudin; RP11-459F3.2; Spastic paraplegia 33 protein; zinc finger FYVE domain containing 27; Zinc finger FYVE domain-containing protein 27; zinc finger, FYVE domain containing 27
Gene Aliases: PROTRUDIN; SPG33; ZFYVE27
UniProt ID: (Human) Q5T4F4
Entrez Gene ID: (Human) 118813
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support