Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Peptide sequence: VGGKDGLMDS TPALTPTESL SSQDLTPGSV EEAEEAEPDE EFKDAIEETH
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 75%; Zebrafish: 82%
This gene encodes a protein with several transmembrane domains, a Rab11-binding domain and a lipid-binding FYVE finger domain. The encoded protein appears to promote neurite formation. A mutation in this gene has been reported to be associated with hereditary spastic paraplegia, however the pathogenicity of the mutation, which may simply represent a polymorphism, is unclear.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protrudin; RP11-459F3.2; Spastic paraplegia 33 protein; zinc finger FYVE domain containing 27; Zinc finger FYVE domain-containing protein 27; zinc finger, FYVE domain containing 27
Gene Aliases: PROTRUDIN; SPG33; ZFYVE27
UniProt ID: (Human) Q5T4F4
Entrez Gene ID: (Human) 118813
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support