Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: NQVHLGLRGE LFGRADPYRP VASPRTDPYA AGAQFPNYSP MNMNMGVNVA AHHGPGAFFR YMRQPIKQEL SCKWIDEAQL SRPKKSCDRT FST
This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. This nuclear protein probably functions as a transcription factor in early stages of left-right body axis formation. Mutations in this gene cause X-linked visceral heterotaxy, which includes congenital heart disease and left-right axis defects in organs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: heterotaxy 1; ZIC; Zic family member 3 (odd-paired homolog, Drosophila); ZIC1; ZIC2; Zinc finger protein 203; Zinc finger protein of the cerebellum 3; Zinc finger protein ZIC 3; ZNF201
Gene Aliases: HTX; HTX1; VACTERLX; ZIC3; ZNF203
UniProt ID: (Human) B2CNW4
Entrez Gene ID: (Human) 7547
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support