Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: IMR32.
Predicted reactivity: Mouse (92%), Rat (93%), Bovine (92%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ZIMP7, also known as ZMIZ2, is a novel PIAS (protein inhibitor of activated signal transducer and activator of transcription)-like protein and a transcriptional coactivator. ZIMP7 is expressed most abundantly in testis. The C-terminal proline-rich domain possesses a significant intrinsic transcriptional activity and this activity is inhibited by the N-terminus in the full-length ZIMP7. ZIMP7 and the related protein ZIMP10 interact with PIAS3 and enhances Androgen Receptor (AR)- mediated transcription. The interaction between ZIMP7 and SWI/SNF complex suggests a possible role for ZIMP7 in chromatin modification.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PIAS-like protein Zimp7; Zinc finger MIZ domain-containing protein 2; zinc finger, MIZ-type containing 2
Gene Aliases: HRIHFB2007; hZIMP7; KIAA1886; NET27; TRAFIP20; ZIMP7; ZMIZ2
UniProt ID: (Human) Q8NF64
Entrez Gene ID: (Human) 83637
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support