Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KFTPLPEGK LKEEIEVMAK SIDFPLTKVY VVEGSKRSSH SNAYFYGFFK NKRIVLFDTL LEEYSVLNKD IQEDSGMEPR NEEEGNSEEI KAKVKNKKQG CKNEEVLAVL GHELGHWKLG HTVKNIIISQ MNSFLCFFLF AVLIGRKELF AAFGFYDSQP TLIGLLIIFQ FIFSPYNEVL SFCLTVLSRR FEFQADAFAK KLGKAKDLYS ALIKLNKDNL GFPVSDWLFS MWHYSHPPLL ERLQALKTMK QH (225-475 aa encoded by BC037283)
This gene encodes a member of the peptidase M48A family. The encoded protein is a zinc metalloproteinase involved in the two step post-translational proteolytic cleavage of carboxy terminal residues of farnesylated prelamin A to form mature lamin A. Mutations in this gene have been associated with mandibuloacral dysplasia and restrictive dermopathy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAAX prenyl protease 1 homolog; FACE-1; Farnesylated proteins-converting enzyme 1; FLJ14968; Prenyl protein-specific endoprotease 1; zinc metallopeptidase STE24 homolog; Zinc metalloproteinase Ste24 homolog
Gene Aliases: FACE-1; FACE1; HGPS; PRO1; STE24; Ste24p; ZMPSTE24
UniProt ID: (Human) O75844
Entrez Gene ID: (Human) 10269
Molecular Function: metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support