Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Recommended positive controls: human ESC.
Predicted reactivity: Mouse (95%), Rhesus Monkey (99%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ZNF198 is a ubiquitously expressed nuclear protein whose physiological role is largely unknown. It consists of 5 N-terminal zinc finger motifs responsible for protein-protein interactions, a central proline-rich domain and C-terminal nuclear localization signal (NLS) and an acidic domain. Reciprocal chromosomal translocation involving ZNF198 and FGFR1 results in the formation of ZNF198/FGFR1 fusion kinase protein. This fusion protein is a ligand-dependent, constitutively active cytoplasmic tyrosine kinase and regulates several STAT transcription factors including STAT 1, 3 and 5. This fusion protein is an oncogenic protein and is associated with an atypical myeloproliferative disease, peripheral blood eosinophilia and T-cell leukemia/lymphoma.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fused in myeloproliferative disorders protein; rearranged in an atypical myeloproliferative disorder; Rearranged in atypical myeloproliferative disorder protein; RP11-264J4.7; Zinc finger MYM-type protein 2; Zinc finger protein 198; zinc finger, MYM-type 2
Gene Aliases: 5830413P05Rik; FIM; MYM; RAMP; SCLL; Zfp198; ZMYM2; ZNF198
UniProt ID: (Human) Q9UBW7, (Mouse) Q3UUZ8
Entrez Gene ID: (Human) 7750, (Mouse) 76007
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support