Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
ZMYND17 contains 1 MYND-type zinc finger. The exact function of ZMYND17 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MSS51 mitochondrial translational activator homolog; Putative protein MSS51 homolog, mitochondrial; Zinc finger MYND domain-containing protein 17; zinc finger, MYND domain containing 17; zinc finger, MYND-type containing 17
Gene Aliases: MSS51; ZMYND17
UniProt ID: (Human) Q4VC12
Entrez Gene ID: (Human) 118490
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support