Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MRAHAGHKR SECGGEWRET PRKQKQHGKA SISPSSGARR TVTPTRKRPY ECKVCGKAFN SPNLFQIHQR THTGKRSYKC REIVRAFTVS SFFRKHGKMH TGEKRYECKY CGKPIDYPSL FQIHVRTHTG EKPYKCKQCG KAFISAGYLR THEIRSHALE KSHQCQECGK KLSCSSSLHR HERTHSGGKL YECQKCAKVF RCPTSLQAHE RAHTGERPYE CNKCGKTFNY PSCFRRHKKT HSGEKPYECT RCGKAFGWCS SLRRHEMTHT GEKPFDCKQC GKVFTFSNYL RLHERTHLAG RSQCFGRRQG DHLSPGV (1-316 aa encoded by BC119680)
Zinc finger proteins (ZNFs), such as ZNF101, bind nucleic acids and perform many key functions, the most important of which is regulating transcription (summary by Bellefroid et al., 1993 ). See ZNF91 for general information on ZNFs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp570I0164; MGC149565; MGC149566; Zinc finger protein 101; zinc finger protein 101 (Y2); zinc finger protein 12; Zinc finger protein HZF12
Gene Aliases: HZF12; ZNF101
UniProt ID: (Human) Q8IZC7
Entrez Gene ID: (Human) 94039
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support