Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: VEGEFLLQSY PDEVWQTDDL IERIQEEENK PSRQTVFIET LIEERGNVPG KTFDVETNPV PSRKIAYKNS LCDSCEKCLT SVSEYISSDG SYARMKADEC SGCGKSL
Two members of the human zinc finger Kruppel family, ZNF 12 (KOX 3) and ZNF 26 (KOX 20), have been localized by somatic cell hybrid analysis and in situ chromosomal hybridization. The presence of individual human zinc finger genes in mouse-human hybrid DNAs was correlated with the presence of specific human chromosomes or regions of chromosomes in the corresponding cell hybrids. Analysis of such mouse-human hybrid DNAs assigned the ZNF 12 (KOX 3) gene to chromosome region 7p.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GIOT-3; gonadotropin inducible transcription repressor 3; Gonadotropin-inducible ovary transcription repressor 3; zinc finger protein 11; Zinc finger protein 12; Zinc finger protein 325; Zinc finger protein KOX3
Gene Aliases: GIOT-3; GIOT3; HZF11; KOX3; ZNF12; ZNF325
UniProt ID: (Human) P17014
Entrez Gene ID: (Human) 7559
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support