Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EIHNGGELCD FMENGEIFSE HSCLNAHMGT ENTGDTYDCD EYGENFPMLH NSAPAGETLS VLNQCRKAFS LPPNVHQRTW IGDKSFEYSD CEEAFVDQSH LQANRITH*
ZNF121 may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 121; zinc finger protein 121 (clone ZHC32); Zinc finger protein 20
Gene Aliases: D19S204; ZHC32; ZNF121; ZNF20
UniProt ID: (Human) P58317
Entrez Gene ID: (Human) 7675
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support