Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: RLCESRRGSK CGETTSQMPN VNINKETFTG AKPHECSFCG RDFIHHSSLN RHMRSHTGQK PNEYQEYEKQ PCKCKAVGKT FSYHHCFRKH ER
ZNF14 contains a zinc finger and a Kruppel-associated box (KRAB) domain. KRAB domain is known to be involved in the transcriptional repression of a number of zinc finger proteins.The protein encoded by this gene contains a zinc finger and a Kruppel-associated box (KRAB) domain. KRAB domain is known to be involved in the transcriptional repression of a number of zinc finger proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GIOT-4; gonadotropin inducible transcription repressor-4; Gonadotropin-inducible ovary transcription repressor 4; Zinc finger protein 14; zinc finger protein 14 (KOX 6); Zinc finger protein KOX6
Gene Aliases: GIOT-4; GIOT4; KOX6; ZNF14
UniProt ID: (Human) P17017
Entrez Gene ID: (Human) 7561
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support