Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MNTNSKEVLS LGVQVPEAWE ELLTMKVEAK SHLQWQESRL KRSNPLAREI FRRHFRQLCY QETPGPREAL TRLQELCYQW LRPHVSTKEQ ILDLLVLEQF LSILPKELQG WVREHCPESG EEAVILLEDL ERELDEPQHE MVAHRHRQEV LCKEMVPLAE QTPLTLQSQP KEPQLTCDSA QKCHSIGETD EVTKTEDREL VLRKDCPKIV EPHGKMFNEQ TWEVSQQDPS HGEVGEHKDR IERQWGNLLG EGQHKCDECG KSFTQSSGLI RHQRIHTGER PYECNECGKA FSRSSGLFNH RGIHNIQKRY HCKECGKVFS QSAGLIQHQR IHKGEKPYQC SQCSKSYSRR SFLIEHQRSH TGERPHQCIE CGKSFNRHCN LIRHQKIHTV AELV
ZNF193 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 5 C2H2-type zinc fingers and 1 SCAN box domain. ZNF193 may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell proliferation-inducing gene 12 protein; cell proliferation-inducing protein 12; PRD51; Zinc finger and SCAN domain-containing protein 9; Zinc finger protein 193
Gene Aliases: PIG12; PRD51; ZNF193; ZSCAN9
UniProt ID: (Human) O15535
Entrez Gene ID: (Human) 7746
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support