Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MNTNSKEVLS LGVQVPEAWE ELLTMKVEAK SHLQWQESRL KRSNPLAREI FRRHFRQLCY QETPGPREAL TRLQELCYQW LRPHVSTKEQ
ZNF193 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 5 C2H2-type zinc fingers and 1 SCAN box domain. ZNF193 may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell proliferation-inducing gene 12 protein; cell proliferation-inducing protein 12; PRD51; Zinc finger and SCAN domain-containing protein 9; Zinc finger protein 193
Gene Aliases: PIG12; PRD51; ZNF193; ZSCAN9
UniProt ID: (Human) O15535
Entrez Gene ID: (Human) 7746
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support