Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 5
This antibody is predicted to react with Bovine.
ZNF207 is a new candidate transcription factor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 49 kDa zinc finger protein; BUB3-interacting and GLEBS motif-containing protein ZNF207; BuGZ; Zinc finger protein 207
Gene Aliases: 8430401D15Rik; BUGZ; hBuGZ; Zep; Zfp207; ZNF207
UniProt ID: (Human) O43670, (Mouse) Q9JMD0
Entrez Gene ID: (Human) 7756, (Mouse) 22680
Molecular Function:
DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support