Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Peptide Sequence: MQPLSKLMAI SKPRNLSLRE QREVLRADMS WQQETNPVVE THDSEASRQK FRHFQYLKVS GPHEALSQLW ELCLQWLRPE IHTKKQIIEL LVLEQFLAIL PEEVRTWVNL QHPNNSKDMV TLIEDVIEML EDEDMPCKDS ALQMGSIKEK MKAGSRTGKP QEPVTFKDVV VEFSKEEWGQ LDSAVKNLYR NVMLENFRNL NSLRKAHLLS KPFESLKLES KKKRWIMEKE IPRKTIFDMK SISGEESSHG VIMTRLTESG HPSSDAWKDG VSLLLPRLEY NDAVSAHCNL HLWVQVILLP QPPE
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BAZ 2; BAZ-2; BWSCR2-associated zinc finger protein 2; Zinc finger protein 215; Zinc finger protein with KRAB and SCAN domains 11
Gene Aliases: BAZ-2; BAZ2; ZKSCAN11; ZNF215; ZSCAN43
UniProt ID: (Human) Q9UL58
Entrez Gene ID: (Human) 7762
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support