Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Binds to the promoters of target genes and functions as repressor. Promotes cell proliferation and antagonizes cell death. Promotes phosphorylation of AKT1 at 'Ser-473'.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 217
Gene Aliases: ZABC1; ZNF217
UniProt ID: (Human) O75362
Entrez Gene ID: (Human) 7764
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support