Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: CNECGKFFSQ SSSLIRHRRS HTGERPYECS ECWKSFSNHS SLVKHRRVHT GERPYECSEC GKSFSQSSNL TNHQRIHSGE RPYECSDCGK FFTFNSNLLK HQNVHKG
Transcriptional repressor that plays a role in cell proliferation. Requires TRIM28 for its activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BMZF-3; Bone marrow zinc finger 3; Zinc finger protein 256
Gene Aliases: BMZF-3; BMZF3; ZNF256
UniProt ID: (Human) Q9Y2P7
Entrez Gene ID: (Human) 10172
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support