Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: PEGNMEDKEM TGPQLPESLE DVAMYISQEE WGHQDPSKRA LSRDTVQESY ENVDSLESHI PSQEVPGTQV GQGGKLWDPS VQSCKEGLSP RGPAPGEE
Might play an important role in basic cellular processes as a transcriptional repressor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 263; Zinc finger protein FPM315; Zinc finger protein with KRAB and SCAN domains 12
Gene Aliases: FPM315; ZKSCAN12; ZNF263; ZSCAN44
UniProt ID: (Human) O14978
Entrez Gene ID: (Human) 10127
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support