Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Peptide sequence: TEHALLGRQP RTPERQKPCA QEVPGRTFGS AQDLEAAGGR GHHRMGAVWQ
Sequence homology: Human: 100%
ZNF324 belongs to the krueppel C2H2-type zinc-finger protein family.It contains 9 C2H2-type zinc fingers and 1 KRAB domain. ZNF324 may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 324A; Zinc finger protein ZF5128
Gene Aliases: ZF5128; ZNF324; ZNF324A
UniProt ID: (Human) O75467
Entrez Gene ID: (Human) 25799
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support