Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAALFLSAP PQAEVTFEDV AVYLSREEWG RLGPAQRGLY RDVMLETYGN LVSLGVGPAG PKPGVISQLE RGDEPWVLDV QGTSGKEHLR VNSPALGTRT EYKELTSQET FGEEDPQGSE PVEACDHISK SEGSLEKLVE QRGPRAVTLT NGESSRESGG NLRLLSRPVP DQRPHKCDIC EQSFEQRSYL NNHKRVHRSK KTNTVRNSGE IFSANLVVKE DQKIPTGKKL HYCSYCGKTF RYSANLVKHQ RLHTEEKPYK CDECGKAFSQ SCEFINHRRM HSGEIPYRCD ECGKTFTRRP NLMKHQRIHT GEKPYKCGEC GKHFSAYSSL IYHQRIHTGE KPYKCNDCGK A (1-350 aa encoded by BC004480)
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 34; zinc finger protein 34 (KOX 32); Zinc finger protein KOX32
Gene Aliases: KOX32; ZNF34
UniProt ID: (Human) Q8IZ26
Entrez Gene ID: (Human) 80778
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support