Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MMVGVALRPP CSHPLGCISW LGGRWVRPPT VLLPLACFCC LPAPTSPLWT TPGLQQSLSF QAEVTFEDVA VYLSREEWGR LGPAQRGLYR DVMLETYGNL VSLALGTRTE YKELTSQETF GEEDPQGSEP VEACDHISKS EGSLEKLVEQ RGPRAVTLTN GESSRESGGN LRLLSRPVPD QRPHKCDICE QSFEQRSYLN NHKRVHRSKK TNTVRNSGEI FSANLVVKED QKIPTGKKLH YCSYCGKTFR YSANLVKHQR LHTEEKPYKC DECGKAFSQS CEFINHRRMH SGEIPYRCDE CGKTFTRRPN LMKHQRIHTG EKPYKCGECG KHFSAYSSLI YHQRIHTGEK PYKCNDCGKA FSDGSILIRH RRTHTGEKPF ECKECGKGFT QSSNLIQHQR IHTGEKPYKC NECEKAFIQK TKLVEHQRSH TGEKPYECND CGKVFSQSTH LIQHQRIHTG EKPYKCSECG KAFHNSSRLI HHQRLHHGEK PYRCSDCKKA FSQSTYLIQH RRIHTGEKPY KCSECGKAFR HSSNMCQHQR IHLREDFSM
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 34; zinc finger protein 34 (KOX 32); Zinc finger protein KOX32
Gene Aliases: KOX32; ZNF34
UniProt ID: (Human) Q8IZ26
Entrez Gene ID: (Human) 80778
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support